Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Neu2
Entrez Gene ID
23956
Gene Name
neuraminidase 2
Gene Aliases
MBS, MSS, MTS
Chromosome Location
Chr.1: 87574027 - 87597845 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160163.1 NP_001153635.1 3 - 4 417 59
NM_001160164.1 NP_001153636.1 2 - 3 315 59
NM_001160165.1 NP_001153637.1 2 - 3 443 59
NM_015750.3 NP_056565.1 2 - 3 283 59
GenBank mRNA AB028023.1 - 2 - 3 285 59
AB048604.1 - 2 - 3 540 59
AF139059.1 - 2 - 3 397 59
AK009828.1 - 1 - 2 497 59
AK145284.1 - 2 - 3 316 59
BC026408.1 - 1 - 2 71 59
BC105657.1 - 2 - 3 285 59
BC105658.1 - 2 - 3 285 59
BC105659.1 - 2 - 3 285 59
BC105661.1 - 1 - 2 545 59

View Full Product Details