Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnliprp1
Entrez Gene ID
18946
Gene Name
pancreatic lipase related protein 1
Gene Aliases
Plrp1
Chromosome Location
Chr.19: 58728887 - 58744169 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018874.2 NP_061362.1 6 - 7 727 69
GenBank mRNA AF061274.1 - 6 - 7 728 69
AK028105.1 - 6 - 7 640 69
BC090985.1 - 6 - 7 639 69

View Full Product Details