The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018874.2 | NP_061362.1 | 12 - 13 | 1500 | 74 |
GenBank mRNA | AF061274.1 | - | 12 - 13 | 1501 | 74 |
AK028105.1 | - | 12 - 13 | 1413 | 74 | |
BC068266.1 | - | 8 - 9 | 1056 | 74 | |
BC090985.1 | - | 12 - 13 | 1412 | 74 |