The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018878.3 | NP_061366.2 | 1 - 2 | 352 | 55 |
GenBank mRNA | AF104261.1 | - | 1 - 2 | 352 | 55 |
AK144606.1 | - | 1 - 2 | 214 | 55 | |
AK148141.1 | - | 1 - 2 | 201 | 55 | |
AK198400.1 | - | 1 - 2 | 23 | 55 | |
BC066014.1 | - | 1 - 2 | 141 | 55 |