The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019540.2 | NP_062413.2 | 1 - 2 | 144 | 72 |
GenBank mRNA | AF250839.1 | - | 1 - 2 | 120 | 72 |
AK141262.1 | - | 1 - 2 | 78 | 72 | |
BC052215.1 | - | 1 - 2 | 112 | 72 | |
CJ075286.1 | - | 1 - 2 | 144 | 72 |