Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap4m1
Entrez Gene ID
11781
Gene Name
adaptor-related protein complex AP-4, mu 1
Gene Aliases
4930443L05Rik, Ap4m4
Chromosome Location
Chr.5: 138172021 - 138178687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021392.4 NP_067367.3 14 - 15 1309 71
GenBank mRNA AF242858.1 - 14 - 15 1177 71
AK031638.1 - 14 - 15 1312 71
BC011174.1 - 14 - 15 1194 71

View Full Product Details