Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efcc1
Entrez Gene ID
58229
Gene Name
EF hand and coiled-coil domain containing 1
Gene Aliases
Ccdc48
Chromosome Location
Chr.6: 87730820 - 87755912 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159697.1 NP_001153169.1 1 - 2 677 93
GenBank mRNA AB041550.1 - 1 - 2 677 93
BC139438.1 - 1 - 2 674 93
BC139439.1 - 1 - 2 673 93

View Full Product Details