Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1a1
Entrez Gene ID
58810
Gene Name
aldo-keto reductase family 1, member A1 (aldehyde reductase)
Gene Aliases
2610201A18Rik, Akr1a4
Chromosome Location
Chr.4: 116636510 - 116651674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021473.3 NP_067448.1 2 - 3 425 96
GenBank mRNA AF225564.1 - 2 - 3 330 96
AK005376.1 - 2 - 3 361 96
AK009462.1 - 2 - 3 202 96
AK011321.1 - 2 - 3 354 96
AK011388.1 - 2 - 3 380 96
AK011667.1 - 2 - 3 381 96
AK011794.1 - 2 - 3 354 96
AK011856.1 - 2 - 3 381 96
AK011906.1 - 2 - 3 359 96
AK011908.1 - 2 - 3 394 96
AK011918.1 - 2 - 3 380 96
AK128906.1 - 2 - 3 147 96
AK144198.1 - 2 - 3 381 96
AK146081.1 - 2 - 3 379 96
AK146278.1 - 2 - 3 381 96
AK152086.1 - 2 - 3 360 96
AK159573.1 - 2 - 3 379 96
AK164461.1 - 2 - 3 380 96
AK168950.1 - 2 - 3 376 96
AK186333.1 - 2 - 3 182 96
AK192030.1 - 2 - 3 183 96
AK210646.1 - 2 - 3 97
AK219186.1 - 2 - 3 182 96
AY207463.1 - 2 - 3 402 96
BC003850.1 - 2 - 3 209 96
BC039926.1 - 2 - 3 374 96
BC046762.1 - 2 - 3 331 96

View Full Product Details