The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021534.3 | NP_067509.1 | 3 - 4 | 414 | 69 |
GenBank mRNA | AJ271476.1 | - | 3 - 4 | 403 | 69 |
AK003127.1 | - | 3 - 4 | 429 | 69 | |
AK019381.1 | - | 3 - 4 | 429 | 69 | |
AK050647.1 | - | 3 - 4 | 415 | 69 | |
AK156743.1 | - | - | 414 | 69 | |
BC025796.1 | - | 3 - 4 | 393 | 69 |