The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022031.2 | NP_071314.1 | 6 - 7 | 981 | 60 |
GenBank mRNA | AB049055.1 | - | 6 - 7 | 910 | 60 |
AK015034.1 | - | 1 - 2 | 76 | 60 | |
BC023911.1 | - | 5 - 6 | 1262 | 60 | |
BC138584.1 | - | 5 - 6 | 866 | 60 | |
BC138585.1 | - | 5 - 6 | 866 | 60 |