Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abhd8
Entrez Gene ID
64296
Gene Name
abhydrolase domain containing 8
Gene Aliases
0910001L24Rik, AB030191
Chromosome Location
Chr.8: 71456700 - 71463657 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022419.3 NP_071864.2 1 - 2 188 94
GenBank mRNA AB030191.1 - 1 - 2 178 94
AK003090.1 - 1 - 2 176 94
AK037669.1 - 1 - 2 154 94
AK080618.1 - 1 - 2 186 94
AK155503.1 - 1 - 2 188 94
AK178916.1 - 1 - 2 78 94
AK212822.1 - 1 - 2 95 94
BC026540.1 - 1 - 2 137 94

View Full Product Details