Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp1r7
Entrez Gene ID
66385
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 7
Gene Aliases
2310014J01Rik, SDS22
Chromosome Location
Chr.1: 93343645 - 93367619 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023200.2 NP_075689.1 7 - 8 732 71
GenBank mRNA AF222867.1 - 7 - 8 732 71
AK009342.1 - 7 - 8 747 71
AK009870.1 - 7 - 8 725 71
AK031983.1 - - 1525 71
AK129024.1 - 7 - 8 749 71
AK145136.1 - 7 - 8 738 71
AK162517.1 - 7 - 8 740 71
BC013524.1 - 7 - 8 733 71

View Full Product Details