Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan13
Entrez Gene ID
66109
Gene Name
tetraspanin 13
Gene Aliases
1100001I23Rik, NET-6, Tm4sf13
Chromosome Location
Chr.12: 36014555 - 36042478 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025359.3 NP_079635.1 3 - 4 480 110
GenBank mRNA AK008175.1 - 3 - 4 420 110
AK012571.1 - 3 - 4 492 110
AK090217.1 - 3 - 4 492 110
AK158761.1 - 3 - 4 485 110
AK165768.1 - 3 - 4 517 110
AK167495.1 - 3 - 4 532 110
AK167979.1 - 3 - 4 485 110
BC018317.1 - 3 - 4 480 110

View Full Product Details