Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcts2
Entrez Gene ID
66405
Gene Name
malignant T cell amplified sequence 2
Gene Aliases
2400002F11Rik
Chromosome Location
Chr.2: 152687148 - 152687946 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025543.2 NP_079819.1 1 - 1 52 59
GenBank mRNA AK002563.1 - 1 - 1 30 59
AK010255.1 - 1 - 1 53 59
AK028001.1 - 1 - 1 31 59
BC027507.1 - 1 - 1 45 59

View Full Product Details