The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025822.3 | NP_080098.1 | 9 - 10 | 1040 | 71 |
GenBank mRNA | AK004742.2 | - | 9 - 10 | 994 | 71 |
AK168539.1 | - | 8 - 9 | 885 | 71 | |
AK169340.1 | - | 9 - 10 | 994 | 71 | |
BC043045.1 | - | 9 - 10 | 1028 | 71 | |
BC083123.1 | - | 9 - 10 | 991 | 71 |