Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plin3
Entrez Gene ID
66905
Gene Name
perilipin 3
Gene Aliases
1300012C15Rik, M6prbp1, Tip47
Chromosome Location
Chr.17: 56278962 - 56290511 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025836.3 NP_080112.1 2 - 3 142 59
GenBank mRNA AK004970.1 - 2 - 3 143 59
AK012517.1 - 2 - 3 143 59
AK049537.1 - 2 - 3 143 59
AK050140.1 - 2 - 3 144 59
AK132313.1 - - 143 59
AK167214.1 - 2 - 3 141 59
BC011116.1 - 2 - 3 138 59
DQ257436.1 - 2 - 3 101 59

View Full Product Details