Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsn1
Entrez Gene ID
66934
Gene Name
DSN1, MIND kinetochore complex component, homolog (S. cerevisiae)
Gene Aliases
1700022L09Rik, AW552447
Chromosome Location
Chr.2: 156995062 - 157007075 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025853.3 NP_080129.2 2 - 3 132 59
GenBank mRNA AK006246.1 - 2 - 3 133 59
AK017813.1 - 2 - 3 87 59
AK139887.1 - 2 - 3 701 59
AK145373.1 - 2 - 3 87 59
BC025079.1 - 2 - 3 115 59
BC046807.1 - 2 - 3 93 59

View Full Product Details