Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dsn1
Entrez Gene ID
66934
Gene Name
DSN1, MIND kinetochore complex component, homolog (S. cerevisiae)
Gene Aliases
1700022L09Rik, AW552447
Chromosome Location
Chr.2: 156995062 - 157007075 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025853.3 NP_080129.2 9 - 10 975 64
GenBank mRNA AK006246.1 - 9 - 10 976 64
AK017740.1 - 9 - 10 976 64
AK017813.1 - 9 - 10 930 64
AK139887.1 - 9 - 10 1545 64
AK145373.1 - 10 - 11 803 64
BC025079.1 - 9 - 10 958 64
BC046807.1 - 9 - 10 936 64

View Full Product Details