The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025998.3 | NP_080274.2 | 3 - 4 | 431 | 64 |
GenBank mRNA | AK011164.1 | - | 3 - 4 | 564 | 64 |
AK011850.1 | - | 3 - 4 | 362 | 64 | |
BC116391.1 | - | 3 - 4 | 361 | 64 | |
BC116392.1 | - | 4 - 4 | 81 | ||
EF058048.2 | - | 3 - 4 | 484 | 64 |