The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026119.3 | NP_080395.1 | 1 - 2 | 137 | 113 |
GenBank mRNA | AK010270.1 | - | 1 - 2 | 139 | 113 |
AK010686.1 | - | 1 - 2 | 139 | 113 | |
AK078288.1 | - | 1 - 2 | 149 | 113 | |
AK145636.1 | - | 1 - 2 | 162 | 113 | |
BC024950.1 | - | 1 - 2 | 137 | 113 |