The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007661.3 | NP_031687.2 | 19 - 20 | 2347 | 56 |
GenBank mRNA | AK077668.1 | - | 19 - 20 | 2348 | 56 |
AK147133.1 | - | 19 - 20 | 2325 | 56 | |
BC025058.1 | - | 8 - 9 | 997 | 56 | |
BC052920.1 | - | 19 - 20 | 2425 | 56 | |
L37092.1 | - | 18 - 19 | 2919 | 56 | |
M58633.1 | - | 14 - 15 | 1690 | 56 |