The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007670.4 | NP_031696.1 | 1 - 2 | 369 | 112 |
GenBank mRNA | AF059567.1 | - | 1 - 2 | 278 | 112 |
AK144132.1 | - | 1 - 2 | 379 | 112 | |
AK146668.1 | - | 1 - 2 | 330 | 112 | |
BC002010.1 | - | 1 - 2 | 283 | 112 | |
DT919921.1 | - | 1 - 2 | 108 | 112 |