Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpd
Entrez Gene ID
12874
Gene Name
carboxypeptidase D
Gene Aliases
AA960140, AW322530, D830034L15Rik
Chromosome Location
Chr.11: 76777208 - 76847008 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007754.2 NP_031780.2 8 - 9 2167 134
GenBank mRNA AK032306.1 - 8 - 9 1659 134
AK134907.1 - 10 - 11 2102 134
AK137737.1 - 8 - 9 2146 134
D85391.1 - 8 - 9 2155 134

View Full Product Details