Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctsk
Entrez Gene ID
13038
Gene Name
cathepsin K
Gene Aliases
AI323530, MMS10-Q, Ms10q, catK
Chromosome Location
Chr.3: 95499210 - 95509387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007802.4 NP_031828.2 4 - 5 526 84
GenBank mRNA AK003425.1 - 4 - 5 451 84
AK132648.1 - 4 - 5 456 84
BC046320.1 - 4 - 5 480 84
X94444.1 - 3 - 4 84

View Full Product Details