Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsl1
Entrez Gene ID
14081
Gene Name
acyl-CoA synthetase long-chain family member 1
Gene Aliases
Acas, Acas1, Acs, FACS, Facl2, LACS 1, LACS1
Chromosome Location
Chr.8: 46471037 - 46536051 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK004897.1 - 17 - 18 1718 120
AK145900.1 - 17 - 18 1845 120
AK149406.1 - 17 - 18 1723 120
AK152772.1 - 17 - 18 1723 120
AK153050.1 - 17 - 18 1851 120
AK161189.1 - 17 - 18 1697 120
AK168078.1 - 17 - 18 1723 120
AK180785.1 - 2 - 3 218 120
BC056644.1 - 17 - 18 1707 120
U15977.1 - 16 - 17 1636 120

View Full Product Details