Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myt1l
Entrez Gene ID
17933
Gene Name
myelin transcription factor 1-like
Gene Aliases
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106
Chromosome Location
Chr.12: 29528311 - 29923216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001093775.1 NP_001087244.1 22 - 23 4079 123
NM_001093776.1 NP_001087245.1 22 - 23 3985 123
NM_001093778.1 NP_001087247.1 22 - 23 4070 123
NM_008666.3 NP_032692.2 22 - 23 4073 123
GenBank mRNA AB093283.1 - 19 - 20 3432 123
AB212898.1 - 21 - 22 3677 123
AF004295.1 - 22 - 23 3889 123
BC079651.1 - 22 - 23 3954 123
BC094438.1 - 18 - 19 3310 123
BC131677.1 - 21 - 22 3674 123
U86338.1 - 22 - 23 3801 123

View Full Product Details