Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rad54l
Entrez Gene ID
19366
Gene Name
RAD54 like (S. cerevisiae)
Gene Aliases
RAD54
Chromosome Location
Chr.4: 116096727 - 116123690 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122958.1 NP_001116430.1 10 - 11 1333 87
NM_001122959.1 NP_001116431.1 10 - 11 1312 87
NM_009015.3 NP_033041.3 9 - 10 1663 87
GenBank mRNA AK030753.1 - 10 - 11 1291 87
AK088876.1 - 10 - 11 1330 87
AK161644.1 - 10 - 11 1286 87
BC021643.1 - 9 - 10 1644 87
X97796.1 - 9 - 10 1123 87

View Full Product Details