The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009060.2 | NP_033086.1 | 3 - 4 | 418 | 72 |
GenBank mRNA | AK146465.1 | - | 3 - 4 | 450 | 72 |
BC012710.1 | - | 3 - 4 | 431 | 72 | |
D86217.1 | - | 2 - 3 | 370 | 72 | |
U28937.1 | - | 3 - 4 | 418 | 72 |