Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgn
Entrez Gene ID
19733
Gene Name
regucalcin
Gene Aliases
AI265316, GNL, SMP30
Chromosome Location
Chr.X: 20549767 - 20562089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009060.2 NP_033086.1 6 - 7 927 102
GenBank mRNA AK146465.1 - 6 - 7 959 102
BC012710.1 - 6 - 7 940 102
D86217.1 - 5 - 6 879 102
U28937.1 - 6 - 7 927 102

View Full Product Details