Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scg3
Entrez Gene ID
20255
Gene Name
secretogranin III
Gene Aliases
1B1075, AI385542, Chgd, SgIII
Chromosome Location
Chr.9: 75643366 - 75684082 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164790.1 NP_001158262.1 2 - 3 473 115
NM_009130.3 NP_033156.1 2 - 3 473 115
GenBank mRNA AK133655.1 - 2 - 3 375 115
AK139140.1 - 2 - 3 413 115
AK148498.1 - 2 - 3 455 115
AK158315.1 - 2 - 3 346 115
AK164089.1 - 2 - 3 416 115
BC024785.1 - 2 - 3 432 115
BU058741.1 - 2 - 3 473 115
U02982.1 - 2 - 3 437 115

View Full Product Details