Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sepw1
Entrez Gene ID
20364
Gene Name
selenoprotein W, muscle 1
Gene Aliases
selW
Chromosome Location
Chr.7: 15917208 - 15922371 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009156.2 NP_033182.1 1 - 2 122 77
GenBank mRNA AF015284.1 - 1 - 2 77 77
AF241527.2 - 1 - 2 75 77
AK002870.1 - 1 - 2 98 77
AK028124.1 - 1 - 2 92 77
BC052719.1 - 1 - 2 84 77
BU604713.1 - 1 - 2 155 77
U67890.1 - 1 - 2 74 77

View Full Product Details