Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sftpd
Entrez Gene ID
20390
Gene Name
surfactant associated protein D
Gene Aliases
AI573415, SP-D, Sftp4
Chromosome Location
Chr.14: 41172212 - 41185198 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009160.2 NP_033186.1 4 - 5 507 61
GenBank mRNA BC003705.1 - 4 - 5 458 61
BC012896.1 - 3 - 4 695 61
L40156.1 - 3 - 4 435 61

View Full Product Details