Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Scg5
- Entrez Gene ID
-
20394
- Gene Name
- secretogranin V
- Gene Aliases
-
7B2, AI325031, Sgne-1, Sgne1
- Chromosome Location
-
Chr.2: 113776313 - 113829091 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X15830.1 |
- |
1 - 2 |
|
95 |
View Full Product Details