Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptbn1
Entrez Gene ID
20742
Gene Name
spectrin beta, non-erythrocytic 1
Gene Aliases
9930031C03Rik, AL033301, SPTB2, Spnb-2, Spnb2, elf1, elf3, mKIAA4049
Chromosome Location
Chr.11: 30099395 - 30268204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009260.2 NP_033286.2 28 - 29 6437 60
NM_175836.2 NP_787030.2 28 - 29 6097 60
GenBank mRNA AF017112.1 - 28 - 29 6344 60
AK051630.1 - 3 - 4 455 60
AK161119.1 - 7 - 8 1220 60
AK220456.1 - 29 - 30 6299 60
M74773.1 - 28 - 29 6417 60

View Full Product Details