The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109873.1 | NP_001103343.1 | 2 - 3 | 465 | 88 |
NM_009824.2 | NP_033954.2 | 2 - 3 | 446 | 88 | |
GenBank mRNA | AF038029.1 | - | 2 - 3 | 328 | 88 |
BB658300.1 | - | 2 - 3 | 208 | 88 | |
BC141396.1 | - | 2 - 3 | 275 | 88 | |
BC151156.1 | - | 2 - 3 | 275 | 88 |