The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109873.1 | NP_001103343.1 | 7 - 8 | 1284 | 94 |
NM_009824.2 | NP_033954.2 | 7 - 8 | 1265 | 94 | |
NM_177289.1 | NP_796263.1 | 6 - 7 | 1206 | 94 | |
GenBank mRNA | AF038029.1 | - | 7 - 8 | 1147 | 94 |
AK033680.1 | - | 6 - 7 | 93 | ||
AK041887.1 | - | 7 - 8 | 1287 | 94 | |
BC141396.1 | - | 7 - 8 | 1094 | 94 | |
BC151156.1 | - | 7 - 8 | 1094 | 94 |