The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009867.2 | NP_033997.1 | 3 - 4 | 541 | 76 |
GenBank mRNA | BC047991.1 | - | 3 - 4 | 541 | 76 |
D14888.1 | - | 3 - 4 | 451 | 76 | |
X69966.1 | - | 3 - 4 | 397 | 76 |