Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift81
Entrez Gene ID
12589
Gene Name
intraflagellar transport 81
Gene Aliases
AW060663, Cdv-1, Cdv-1r, Cdv1
Chromosome Location
Chr.5: 122540265 - 122614546 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009879.3 NP_034009.2 12 - 13 1438 83
GenBank mRNA AF354757.1 - 12 - 13 1438 83
BC055721.1 - 12 - 13 1395 83
Y10495.2 - 12 - 13 1438 83

View Full Product Details