The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010779.2 | NP_034909.2 | 3 - 4 | 380 | 117 |
GenBank mRNA | AB051900.1 | - | 3 - 4 | 328 | 117 |
AK144385.1 | - | 3 - 4 | 381 | 117 | |
AY007569.1 | - | 3 - 4 | 380 | 117 | |
BC026198.1 | - | 3 - 4 | 356 | 117 | |
CJ046459.1 | - | 3 - 4 | 383 | 117 | |
M55617.1 | - | - | 117 | ||
X68804.1 | - | 3 - 4 | 327 | 117 |