Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tpsb2
- Entrez Gene ID
-
17229
- Gene Name
- tryptase beta 2
- Gene Aliases
-
AV011504, MMCP-6, Mcp-6, Mcp6, Mcpt6
- Chromosome Location
-
Chr.17: 25366303 - 25368096 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X78542.1 |
- |
- |
675 |
109 |
View Full Product Details