The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136072.2 | NP_001129544.1 | 1 - 2 | 257 | 66 |
NM_001159567.1 | NP_001153039.1 | 1 - 2 | 257 | 66 | |
NM_001159568.1 | NP_001153040.1 | 1 - 2 | 257 | 66 | |
NM_010825.3 | NP_034955.1 | 1 - 2 | 257 | 66 | |
GenBank mRNA | AJ000504.1 | - | 1 - 2 | 246 | 66 |
AJ000505.1 | - | 1 - 2 | 246 | 66 | |
AJ000506.1 | - | 1 - 2 | 246 | 66 | |
AJ000507.1 | - | 1 - 2 | 246 | 66 | |
AK146633.1 | - | 1 - 2 | 257 | 66 | |
BC017375.1 | - | 1 - 2 | 323 | 66 | |
U57343.1 | - | - | 66 | ||
U68383.1 | - | 1 - 2 | 216 | 66 | |
U68384.1 | - | 1 - 2 | 216 | 66 |