The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010850.2 | NP_034980.2 | 1 - 1 | 889 | 102 |
GenBank mRNA | AK132789.1 | - | 1 - 1 | 890 | 102 |
BC117074.1 | - | 1 - 1 | 668 | 102 | |
BC120777.1 | - | 1 - 1 | 668 | 102 |