The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110350.1 | NP_001103820.1 | 14 - 15 | 2568 | 72 |
NM_001110351.1 | NP_001103821.1 | 15 - 16 | 2445 | 72 | |
NM_011378.2 | NP_035508.2 | 15 - 16 | 2676 | 72 | |
GenBank mRNA | AK032209.1 | - | 14 - 15 | 2436 | 72 |
AK136347.1 | - | 9 - 10 | 1370 | 72 | |
AK220292.1 | - | 14 - 15 | 2545 | 72 | |
BC052716.1 | - | 14 - 15 | 2455 | 72 | |
BC053385.1 | - | 14 - 15 | 2438 | 72 | |
BC078454.1 | - | 14 - 15 | 2429 | 72 | |
L36831.1 | - | 14 - 15 | 2359 | 72 | |
L38620.2 | - | 13 - 14 | 2335 | 72 | |
L38621.2 | - | 13 - 14 | 2335 | 72 | |
U22394.1 | - | 15 - 16 | 2684 | 72 |