The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011484.2 | NP_035614.1 | 6 - 7 | 712 | 72 |
GenBank mRNA | AK035924.1 | - | 6 - 7 | 635 | 72 |
AK144987.1 | - | 6 - 7 | 729 | 72 | |
AK147832.1 | - | 6 - 7 | 622 | 72 | |
AK157767.1 | - | 6 - 7 | 659 | 72 | |
AK162431.1 | - | 6 - 7 | 660 | 72 | |
AK163922.1 | - | 6 - 7 | 649 | 72 | |
AK169170.1 | - | 6 - 7 | 719 | 72 | |
BC044666.1 | - | 6 - 7 | 712 | 72 | |
BC055326.1 | - | 6 - 7 | 658 | 72 | |
U43900.1 | - | 6 - 7 | 569 | 72 |