The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253897.1 | NP_001240826.1 | 4 - 5 | 789 | 80 |
NM_001253898.1 | NP_001240827.1 | 3 - 4 | 611 | 80 | |
NM_012029.2 | NP_036159.1 | 4 - 5 | 797 | 80 | |
GenBank mRNA | AF182510.1 | - | 3 - 4 | 810 | 80 |
BC004583.1 | - | 3 - 4 | 598 | 80 | |
BC016424.1 | - | 4 - 5 | 737 | 80 | |
CB589832.1 | - | 2 - 3 | 493 | 80 |