The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253897.1 | NP_001240826.1 | 7 - 8 | 1218 | 109 |
NM_001253898.1 | NP_001240827.1 | 6 - 7 | 1040 | 109 | |
NM_012029.2 | NP_036159.1 | 7 - 8 | 1226 | 109 | |
GenBank mRNA | AF182510.1 | - | 6 - 7 | 1239 | 109 |
AK007564.1 | - | 7 - 8 | 1192 | 109 | |
AK012835.1 | - | 7 - 8 | 1191 | 109 | |
BC004583.1 | - | 6 - 7 | 1027 | 109 | |
BC016424.1 | - | 7 - 8 | 1166 | 109 |