Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mef2a
Entrez Gene ID
17258
Gene Name
myocyte enhancer factor 2A
Gene Aliases
A430079H05Rik
Chromosome Location
Chr.7: 67231163 - 67373501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033713.2 NP_001028885.1 8 - 9 1200 73
GenBank mRNA BC061128.1 - 8 - 9 1171 73
U30823.1 - 6 - 7 949 73

View Full Product Details