Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snap91
Entrez Gene ID
20616
Gene Name
synaptosomal-associated protein 91
Gene Aliases
91kDa, AP180, F1-20, mKIAA0656
Chromosome Location
Chr.9: 86765923 - 86880708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277982.1 NP_001264911.1 5 - 6 560 59
NM_001277983.1 NP_001264912.1 5 - 6 672 59
NM_001277985.1 NP_001264914.1 5 - 6 672 59
NM_001277986.1 NP_001264915.1 5 - 6 672 59
NM_013669.2 NP_038697.1 5 - 6 672 59
NR_102729.1 5 - 6 672 59
GenBank mRNA AK051176.1 - 5 - 6 667 59
AK147087.1 - 5 - 6 663 59
AK159570.1 - 5 - 6 638 59
BC031773.1 - 5 - 6 560 59
BC052454.1 - 5 - 6 672 59
M83985.1 - 5 - 6 597 59

View Full Product Details