The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160404.1 | NP_001153876.1 | 12 - 13 | 1784 | 64 |
NM_013814.3 | NP_038842.3 | 12 - 13 | 1872 | 64 | |
GenBank mRNA | AK083391.1 | - | 12 - 13 | 1788 | 64 |
AK148550.1 | - | 6 - 7 | 852 | 64 | |
BC056215.1 | - | 12 - 13 | 1789 | 64 | |
BC090962.1 | - | 11 - 12 | 1662 | 64 | |
U73820.1 | - | 10 - 11 | 1537 | 64 |