The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045959.1 | NP_001039424.1 | 2 - 3 | 308 | 68 |
NM_001045964.1 | NP_001039429.1 | 2 - 3 | 308 | 68 | |
NM_016713.2 | NP_057922.2 | 2 - 3 | 308 | 68 | |
NM_176893.2 | NP_795712.2 | 2 - 3 | 308 | 68 | |
GenBank mRNA | AB035697.1 | - | 2 - 3 | 293 | 68 |
AB041925.1 | - | 2 - 3 | 294 | 68 | |
BC052474.1 | - | 2 - 3 | 172 | 68 | |
BC079549.1 | - | 2 - 3 | 166 | 68 | |
CB245214.1 | - | 2 - 3 | 172 | 68 |